3-Cyclohexene-1-methanol ≥98.0% (by GC)

Supplier: TCI America

C2435-250G C2435-25G
TCC2435-250GEA 203.82 CAD
TCC2435-250G TCC2435-25G
3-Cyclohexene-1-methanol ≥98.0% (by GC)
3-Cyclohexene-1-methanol

CAS Number: 1679-51-2
MDL Number: MFCD00001574
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 73
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 1,2,3,6-Tetrahydrobenzyl Alcohol

Formula: C₇H₁₂O
Boiling Pt: 100 °C
Density: 0.97 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00001574
CAS Number: 1679-51-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cb069f03281bf3b32d920533cb41b39db1dfddb&vid=4cb069f03281bf3b32d920533cb41b39db1dfddb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR