2-Chloro-6-methyl-5-nitropyridine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
6-Chloro-2-methyl-3-nitropyridine, 2-Chloro-5-nitro-6-picoline, 6-Chloro-3-nitro-2-picoline
C2434-1G
C2434-5G
TCC2434-1GEA
61.62
CAD
TCC2434-1G
TCC2434-5G
2-Chloro-6-methyl-5-nitropyridine ≥98.0% (by GC)
2-Chloro-6-methyl-5-nitropyridine
CAS Number: 22280-60-0
MDL Number: MFCD00234160
Molecular Formula: C6H5ClN2O2
Molecular Weight: 172.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 55
Other Product Names / Synonyms: 6-Chloro-3-nitro-2-picoline
Formula:
C₆H₅ClN₂O₂ MW: 172.57 g/mol Melting Pt: 55 °C |
MDL Number:
MFCD00234160 CAS Number: 22280-60-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...