3-(2-Chloroethyl)-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one ≥98.0%

Supplier: TCI America

Synonyms: 3-(2-Chloroethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one

C2433-25G C2433-5G
TCC2433-25GEA 557.11 CAD
TCC2433-25G TCC2433-5G
3-(2-Chloroethyl)-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one ≥98.0%
3-(2-Chloroethyl)-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

CAS Number: 63234-80-0
MDL Number: MFCD06200813
Molecular Formula: C11H15ClN2O
Molecular Weight: 226.70
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 78

Formula: C₁₁H₁₅ClN₂O
Melting Pt: 78 °C
MDL Number: MFCD06200813
CAS Number: 63234-80-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7627d733828f845c9b50c5084738eed0233b60a&vid=b7627d733828f845c9b50c5084738eed0233b60a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR