Cesium acetate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Acetic acid cesium salt
C2430-100G
C2430-25G
TCC2430-100GEA
235.42
CAD
TCC2430-100G
TCC2430-25G
Cesium acetate ≥98.0% (by titrimetric analysis)
Cesium acetate
CAS Number: 3396-11-0
MDL Number: MFCD00013056
Molecular Formula: C2H3CsO2
Molecular Weight: 191.95
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 193
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: Acetic Acid Cesium Salt
Formula:
H₃CCOOCs MW: 191.95 g/mol Melting Pt: 193 °C |
MDL Number:
MFCD00013056 CAS Number: 3396-11-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...