Copper(II)-2,3,9,10,16,17,23,24-octafluorophthalocyanine ≥98.0% (by total nitrogen and titration analysis), purified by sublimation
Supplier: TCI America
C2427-100MG
C2427-1G
TCC2427-100MGEA
204.93
CAD
TCC2427-100MG
TCC2427-1G
Copper(II)-2,3,9,10,16,17,23,24-octafluorophthalocyanine ≥98.0% (by total nitrogen and titration analysis), purified by sublimation
Copper(II)-2,3,9,10,16,17,23,24-octafluorophthalocyanine
CAS Number: 148651-60-9
Molecular Formula: C32H8CuF8N8
Molecular Weight: 720.01
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Product Categories: Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Highly Purified Reagents, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 2,3,9,10,16,17,23,24-Octafluorophthalocyanine Copper(II), F8CuPc
Formula:
C₃₂H₈CuF₈N₈ |
CAS Number:
148651-60-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...