Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Chloro[1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]copper(I), [1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]chlorocopper(I)

C2422-1G C2422-200MG
TCC2422-1GEA 277.76 CAD
TCC2422-1G TCC2422-200MG
Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I) ≥97.0% (by titrimetric analysis)
Chloro(1,3-dimesitylimidazol-2-ylidene)copper(I)

CAS Number: 873779-78-3
Molecular Formula: C21H24ClCuN2
Molecular Weight: 403.43
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 277
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: [1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]chlorocopper(I)

Formula: C₂₁H₂₄ClCuN₂
MW: 403.43 g/mol
Melting Pt: 277 °C
MDL Number: MFCD15144833
CAS Number: 873779-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cefa29b17ea6234fd47aa3f8c505735076c4464c&vid=cefa29b17ea6234fd47aa3f8c505735076c4464c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR