1,2,4,5-Cyclohexanetetracarboxylic dianhydride ≥98.0%

Supplier: TCI America

Synonyms: HPMDA, 1,2,4,5-Cyclohexanetetracarboxylic Dianhydride

C2419-25G C2419-5G
TCC2419-25GEA 1104.42 CAD
TCC2419-25G TCC2419-5G
1,2,4,5-Cyclohexanetetracarboxylic dianhydride ≥98.0%
1,2,4,5-Cyclohexanetetracarboxylic dianhydride

CAS Number: 2754-41-8
MDL Number: MFCD13704912
Molecular Formula: C10H8O6
Molecular Weight: 224.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 240

Formula: C₁₀H₈O₆
MW: 224.17 g/mol
Melting Pt: 240 °C
MDL Number: MFCD13704912
CAS Number: 2754-41-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc9cd741333dbef7459be0cca0076f678ef4e24&vid=cbc9cd741333dbef7459be0cca0076f678ef4e24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR