2-Cyanophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
C2416-1G
C2416-5G
TCC2416-1GEA
66.04
CAD
TCC2416-1G
TCC2416-5G
2-Cyanophenylboronic acid (contains varying amounts of Anhydride)
2-Cyanophenylboronic acid
CAS Number: 138642-62-3
MDL Number: MFCD01632208
Molecular Formula: C7H6BNO2
Molecular Weight: 146.94
Form: Crystal
Color: White
Melting point (°C): 240
Other Product Names / Synonyms: 2-Cyanobenzeneboronic Acid
Formula:
C₇H₆BNO₂ Melting Pt: 240 °C |
MDL Number:
MFCD01632208 CAS Number: 138642-62-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...