6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

C2404-1G C2404-200MG
TCC2404-1GEA 775.15 CAD
TCC2404-1G TCC2404-200MG
6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one ≥95.0% (by HPLC, total nitrogen)
6-Cyano-9-(diethylamino)-5H-benzo[a]phenoxazin-5-one

CAS Number: 128119-95-9
Molecular Formula: C21H17N3O2
Molecular Weight: 343.39
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal

Formula: C₂₁H₁₇N₃O₂
CAS Number: 128119-95-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3db31d2f648e38fa16c121958c0a5f08e9e0303&vid=a3db31d2f648e38fa16c121958c0a5f08e9e0303&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR