ɑ-Cyclopentyl-DL-mandelic acid ≥98.0%

Supplier: TCI America

Synonyms: alpha-Cyclopentylmandelic Acid, α-Cyclopentylmandelic acid

C2394-25G C2394-5G
TCC2394-25GEA 317.58 CAD
TCC2394-25G TCC2394-5G
ɑ-Cyclopentyl-DL-mandelic acid ≥98.0%
alpha-Cyclopentyl-DL-mandelic acid

CAS Number: 427-49-6
MDL Number: MFCD00019296
Molecular Formula: C13H16O3
Molecular Weight: 220.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 148

Formula: C₁₃H₁₆O₃
Melting Pt: 148 °C
MDL Number: MFCD00019296
CAS Number: 427-49-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5725bbfcb373d042c62e3616777bafb4569e150&vid=b5725bbfcb373d042c62e3616777bafb4569e150&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR