ɑ-Cyclohexyl-DL-mandelic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2393-25G C2393-5G
TCC2393-25GEA 338.12 CAD
TCC2393-25G TCC2393-5G
ɑ-Cyclohexyl-DL-mandelic acid ≥98.0% (by HPLC, titration analysis)
alpha-Cyclohexyl-DL-mandelic acid

CAS Number: 4335-77-7
MDL Number: MFCD00194543
Molecular Formula: C14H18O3
Molecular Weight: 234.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 167

Formula: C₁₄H₁₈O₃
Boiling Pt: 135 °C
Melting Pt: 167 °C
MDL Number: MFCD00194543
CAS Number: 4335-77-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c1c5a58df3a1e90771044acad074b4cec824707&vid=6c1c5a58df3a1e90771044acad074b4cec824707&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR