6-Chloro-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) for biochemical research
Supplier: TCI America
Synonyms:
Rose-Gal, Salmon-Gal
C2371-100MG
C2371-20MG
TCC2371-100MGEA
120.71
CAD
TCC2371-100MG
TCC2371-20MG
6-Chloro-3-indolyl-β-D-galactopyranoside ≥98.0% (by HPLC) for biochemical research
6-Chloro-3-indolyl-beta-D-galactopyranoside
[for Biochemical Research]
CAS Number: 138182-21-5
MDL Number: MFCD00467206
Molecular Formula: C14H16ClNO6
Molecular Weight: 329.73
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -45 deg (C=1, MeOH)
Storage Temperature: <0°C
Product Categories: Bioscience, Galactose, ƒÀ-Galactosidase Substrates
Other Product Names / Synonyms: Salmon-Gal, Rose-Gal
Formula:
C₁₄H₁₆ClNO₆ MW: 329.733 g/mol |
MDL Number:
MFCD00467206 CAS Number: 138182-21-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...