Cyclohexyl-p-toluenesulfonate ≥98.0%
Supplier: TCI America
Synonyms:
Cyclohexyl 4-methylbenzenesulphonate, p-Toluenesulfonic acid Cyclohexyl Ester, Cyclohexyl Tosylate, p-Toluenesulphonic acid Cyclohexyl Ester, Cyclohexyl 4-methylbenzenesulfonate
CAS Number: 953-91-3
MDL Number: MFCD00014291
Molecular Formula: C13H18O3S
Molecular Weight: 254.34
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 45
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: p-Toluenesulfonic Acid Cyclohexyl Ester, Cyclohexyl Tosylate
Formula:
C₁₃H₁₈O₃S MW: 254.34 g/mol Melting Pt: 45 °C |
MDL Number:
MFCD00014291 CAS Number: 953-91-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...