Copper(I)-2-thiophenecarboxylate
Supplier: TCI America
Synonyms:
(2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC
C2312-1G
C2312-5G
TCC2312-1GEA
31.6
CAD
TCC2312-1G
TCC2312-5G
Copper(I)-2-thiophenecarboxylate
Copper(I)-2-thiophenecarboxylate
CAS Number: 68986-76-5
MDL Number: MFCD02183524
Molecular Formula: C5H3CuO2S
Molecular Weight: 190.68
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: 2-Thiophenecarboxylic Acid Copper(I) Salt, (2-Thiophenecarboxylato)copper(I), CuTC
Formula:
C₅H₃CuO₂S MW: 190.68 g/mol |
MDL Number:
MFCD02183524 CAS Number: 68986-76-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...