4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
C2309-1G
C2309-5G
TCC2309-1GEA
76
CAD
TCC2309-1G
TCC2309-5G
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide ≥98.0% (by HPLC, total nitrogen)
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
CAS Number: 41859-57-8
MDL Number: MFCD00218513
Molecular Formula: C15H14ClNO2
Molecular Weight: 275.73
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 174
Other Product Names / Synonyms: N-(4-Chlorobenzoyl)tyramine, 4-Chloro-N-(4-hydroxyphenethyl)benzamide
Formula:
C₁₅H₁₄ClNO₂ Melting Pt: 174 °C |
MDL Number:
MFCD00218513 CAS Number: 41859-57-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...