2-(2-Chlorophenyl)-4,5-diphenylimidazole 98,0
Supplier: TCI America
Synonyms:
2-(2-Chlorophenyl)-4,5-diphenyl-1H-imidazole
C2240-25G
C2240-5G
TCC2240-25GEA
256.28
CAD
TCC2240-25G
TCC2240-5G
2-(2-Chlorophenyl)-4,5-diphenylimidazole 98,0
2-(2-Chlorophenyl)-4,5-diphenylimidazole
CAS Number: 1707-67-1
MDL Number: MFCD00030328
Molecular Formula: C21H15ClN2
Molecular Weight: 330.82
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 197
Formula:
C₂₁H₁₅ClN₂ MW: 330.82 g/mol Melting Pt: 197 °C |
MDL Number:
MFCD00030328 CAS Number: 1707-67-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...