Cesium chloride ≥99.0%

Supplier: TCI America

Synonyms: Cesium Chloride

C2203-100G C2203-25G
TCC2203-100GEA 185.1 CAD
TCC2203-100G TCC2203-25G
Cesium chloride ≥99.0%
Cesium chloride

CAS Number: 7647-17-8
MDL Number: MFCD00010955
Molecular Formula: ClCs
Molecular Weight: 168.36
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 645

Formula: CsCl
Melting Pt: 645 °C
MDL Number: MFCD00010955
CAS Number: 7647-17-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=693b52133ae7d0e231d760410da33d4ce8cb365b&vid=693b52133ae7d0e231d760410da33d4ce8cb365b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR