Cesium bromide ≥99.0%

Supplier: TCI America

Synonyms: Cesiumbromide

C2202-25G C2202-100G
TCC2202-25GEA 105.86 CAD
TCC2202-25G TCC2202-100G
Cesium bromide ≥99.0%
Cesium bromide

CAS Number: 7787-69-1
MDL Number: MFCD00010954
Molecular Formula: BrCs
Molecular Weight: 212.81
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 636

Formula: BrCs
MW: 212.81
Boiling Pt: 1300
Melting Pt: 636
Density: 4.46
MDL Number: MFCD00010954
CAS Number: 7787-69-1
Merck Index: 13,02019
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d56dea47e1fb407a8c8a57525f35648e10e9dab&vid=5d56dea47e1fb407a8c8a57525f35648e10e9dab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR