2-(2-Chlorophenyl)ethanol 97,0

Supplier: TCI America

C2169-1G C2169-5G
TCC2169-1GEA 36.34 CAD
TCC2169-1G TCC2169-5G
2-(2-Chlorophenyl)ethanol 97,0
2-(2-Chlorophenyl)ethanol

CAS Number: 19819-95-5
MDL Number: MFCD00002888
Molecular Formula: C8H9ClO
Molecular Weight: 156.61
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 113
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: 2-Chlorophenethyl Alcohol

Formula: C₈H₉ClO
MW: 156.61
Boiling Pt: 130 °C
Density: 1.19 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00002888
CAS Number: 19819-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9aaa1fbb1c02191a41b3133d54c8678dd68d56c&vid=e9aaa1fbb1c02191a41b3133d54c8678dd68d56c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR