2-Chloro-5-iodobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2168-25G C2168-5G
TCC2168-25GEA 340.49 CAD
TCC2168-25G TCC2168-5G
2-Chloro-5-iodobenzoic acid ≥97.0% (by GC, titration analysis)
2-Chloro-5-iodobenzoic acid

CAS Number: 19094-56-5
MDL Number: MFCD00079731
Molecular Formula: C7H4ClIO2
Molecular Weight: 282.46
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 160

Formula: C₇H₄ClIO₂
Melting Pt: 160 °C
MDL Number: MFCD00079731
CAS Number: 19094-56-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8109800504f0127b6f34e3201b2796cd1f68b7c&vid=b8109800504f0127b6f34e3201b2796cd1f68b7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR