3-Chloro-2-fluorobenzaldehyde ≥95.0%

Supplier: TCI America

C2127-25G C2127-5G
TCC2127-25GEA 472.74 CAD
TCC2127-25G TCC2127-5G
3-Chloro-2-fluorobenzaldehyde ≥95.0%
3-Chloro-2-fluorobenzaldehyde

CAS Number: 85070-48-0
MDL Number: MFCD01631571
Molecular Formula: C7H4ClFO
Molecular Weight: 158.56
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 86
Specific Gravity (20/20): 1.36

Formula: C₇H₄ClFO
Boiling Pt: 214 °C
Density: 1.36 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD01631571
CAS Number: 85070-48-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48304cd21fb753362798176698362d9054cec51c&vid=48304cd21fb753362798176698362d9054cec51c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR