1,3,5-Cyclohexanetricarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C2029-25G
C2029-5G
TCC2029-25GEA
844.67
CAD
TCC2029-25G
TCC2029-5G
1,3,5-Cyclohexanetricarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
1,3,5-Cyclohexanetricarboxylic acid (cis and trans mixture)
CAS Number: 25357-95-3
MDL Number: MFCD00134148
Molecular Formula: C9H12O6
Molecular Weight: 216.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 207
Formula:
C₉H₁₂O₆ Melting Pt: 207 °C |
MDL Number:
MFCD00134148 CAS Number: 25357-95-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...