5-Cyanophthalide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C2001-25G C2001-5G
TCC2001-25GEA 163.69 CAD
TCC2001-25G TCC2001-5G
5-Cyanophthalide ≥98.0% (by HPLC, total nitrogen)
5-Cyanophthalide

CAS Number: 82104-74-3
Molecular Formula: C9H5NO2
Molecular Weight: 159.14
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 204

Formula: C₉H₅NO₂
Melting Pt: 204 °C
MDL Number: MFCD01072882
CAS Number: 82104-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0910e87e182e632733f82f0ec39dd0192759fba&vid=b0910e87e182e632733f82f0ec39dd0192759fba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR