2-Chloro-4'-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America

C1977-25G C1977-5G
TCC1977-25GEA 105.86 CAD
TCC1977-25G TCC1977-5G
2-Chloro-4'-fluoroacetophenone ≥98.0% (by GC)
2-Chloro-4'-fluoroacetophenone

CAS Number: 456-04-2
Molecular Formula: C8H6ClFO
Molecular Weight: 172.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 247
Melting point (°C): 50
Flash Point (°C): 110

Formula: C₈H₆ClFO
Boiling Pt: 247 °C
Melting Pt: 50 °C
Flash Pt: 110 °C
MDL Number: MFCD00011652
CAS Number: 456-04-2
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21b8ca928a842051db8048c0c2142bc17de98226&vid=21b8ca928a842051db8048c0c2142bc17de98226&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR