3-Chloro-N-methylaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C1891-25G C1891-5G
TCC1891-25GEA 737.86 CAD
TCC1891-25G TCC1891-5G
3-Chloro-N-methylaniline ≥97.0% (by GC, titration analysis)
3-Chloro-N-methylaniline

CAS Number: 7006-52-2
MDL Number: MFCD00052014
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 110
Specific Gravity (20/20): 1.16

Formula: C₇H₈ClN
Boiling Pt: 235 °C
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00052014
CAS Number: 7006-52-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3b583a963a788cb08e7e30d4b25b117624eca68&vid=b3b583a963a788cb08e7e30d4b25b117624eca68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR