cis-1,4-Cyclohexanediamine dihydrochloride ≥98.0%

Supplier: TCI America

Synonyms: cis-1,4-Diaminocyclohexane Dihydrochloride

C1799-25G C1799-5G
TCC1799-25GEA 663.6 CAD
TCC1799-25G TCC1799-5G
cis-1,4-Cyclohexanediamine dihydrochloride ≥98.0%
cis-1,4-Cyclohexanediamine dihydrochloride

CAS Number: 2121-79-1
MDL Number: MFCD20487918
Molecular Formula: C6H14N2
Molecular Weight: 187.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: cis-1,4-Diaminocyclohexane Dihydrochloride

Formula: C₆H₁₄N₂
MW: 187.11 g/mol
MDL Number: MFCD20487918
CAS Number: 2121-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93b9456844c70a9763b295dd336256453a724512&vid=93b9456844c70a9763b295dd336256453a724512&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR