2-Chloro-4-fluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C1350-25G C1350-5G
TCC1350-25GEA 213.3 CAD
TCC1350-25G TCC1350-5G
2-Chloro-4-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-4-fluorobenzoic acid

CAS Number: 2252-51-9
MDL Number: MFCD00010615
Molecular Formula: C7H4ClFO2
Molecular Weight: 174.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 185

Formula: C₇H₄ClFO₂
Melting Pt: 185 °C
MDL Number: MFCD00010615
CAS Number: 2252-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b38f82771cc6e10c2777804a5b886a54be648d4d&vid=688f5dce90af5ce8a3616cb07101e805b685494e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR