Chloroiodomethane ≥97.0% (by GC) stabilized

Supplier: TCI America

C1179-100G C1179-25G
TCC1179-100GEA 271.76 CAD
TCC1179-100G TCC1179-25G
Chloroiodomethane ≥97.0% (by GC) stabilized
Chloroiodomethane

(stabilized with Copper chip)
CAS Number: 593-71-5
MDL Number: MFCD00001078
Molecular Formula: CH2ClI
Molecular Weight: 176.38
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Specific Gravity (20/20): 2.44

Formula: CH₂ClI
Boiling Pt: 108 °C
Density: 2.44 (20 °C)
MDL Number: MFCD00001078
CAS Number: 593-71-5
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b080d6aae0a1221dba257d8ee84c4fa8544f398f&vid=b080d6aae0a1221dba257d8ee84c4fa8544f398f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR