1-Chloro-4-iodobenzene ≥99.0% (by GC)

Supplier: TCI America

C1075-250G C1075-25G
TCC1075-250GEA 546.68 CAD
TCC1075-250G TCC1075-25G
1-Chloro-4-iodobenzene ≥99.0% (by GC)
1-Chloro-4-iodobenzene

CAS Number: 637-87-6
MDL Number: MFCD00001053
Molecular Formula: C6H4ClI
Molecular Weight: 238.45
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 227
Melting point (°C): 56
Flash Point (°C): 108


Other Product Names / Synonyms: 4-Chloroiodobenzene

Formula: C₆H₄ClI
Boiling Pt: 227 °C
Melting Pt: 56 °C
Flash Pt: 108 °C
MDL Number: MFCD00001053
CAS Number: 637-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb6ea96ca0c38b31a6694987568988dd178abcdb&vid=dacd91d36703947daa3b2d7ae80316eb79569fed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR