4-Chloro-m-phenylenediamine ≥98.0%

Supplier: TCI America

Synonyms: 1-Chloro-2,4-diaminobenzene, 4-Chlorobenzene-1,3-diamine, 4-Chloro-1,3-diaminobenzene, 4-Chloro-1,3-phenylenediamine

C0710-25G C0710-5G
TCC0710-25GEA 344.44 CAD
TCC0710-25G TCC0710-5G
4-Chloro-m-phenylenediamine ≥98.0%
4-Chloro-m-phenylenediamine

CAS Number: 5131-60-2
MDL Number: MFCD00025284
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 90


Other Product Names / Synonyms: 2,4-Diaminochlorobenzene, 1,3-Diamino-4-chlorobenzene

Formula: C₆H₇ClN₂
Melting Pt: 90 °C
MDL Number: MFCD00025284
CAS Number: 5131-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ab183a6ffc66bf2fbec4388b07299642699eabf&vid=9ab183a6ffc66bf2fbec4388b07299642699eabf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR