1-Chlorotetradecane ≥98.0% (by GC)

Supplier: TCI America

C0622-25ML C0622-500ML
TCC0622-25MLEA 50.56 CAD
TCC0622-25ML TCC0622-500ML
1-Chlorotetradecane ≥98.0% (by GC)
1-Chlorotetradecane

CAS Number: 2425-54-9
MDL Number: MFCD00000958
Molecular Formula: C14H29Cl
Molecular Weight: 232.84
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 152
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Myristyl Chloride, Tetradecyl Chloride

Formula: C₁₄H₂₉Cl
Boiling Pt: 155 °C
Density: 0.87 (20 °C)
Flash Pt: 152 °C
MDL Number: MFCD00000958
CAS Number: 2425-54-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e37ca8a7867f78f995feb6e28d93b7d9742eff08&vid=1ae727524da0024484fda2c148084a1a74fb9ef2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR