1-Chlorooctadecane ≥98.0% (by GC)

Supplier: TCI America

C0235-25ML C0235-500ML
TCC0235-25MLEA 64.73 CAD
TCC0235-25ML TCC0235-500ML
1-Chlorooctadecane ≥98.0% (by GC)
1-Chlorooctadecane

CAS Number: 3386-33-2
MDL Number: MFCD00000960
Molecular Formula: C18H37Cl
Molecular Weight: 288.94
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 184
Freezing point (°C): 20
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Octadecyl Chloride, Stearyl Chloride

Formula: C₁₈H₃₇Cl
Boiling Pt: 158 °C
Density: 0.85 (20 °C)
Flash Pt: 184 °C
MDL Number: MFCD00000960
CAS Number: 3386-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d0c0e9d3099e6f5801a2eb0ae1d2a979981b78b&vid=85dff72f04257f2016ceac7e576aa7c4f4dd8161&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR