3-Bromo-2-hydroxy-5-nitropyridine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Bromo-5-nitro-2(1H)-pyridinone, 3-Bromo-5-nitro-2-pyridinol, 3-Bromo-5-nitro-2-pyridone

B4672-25G B4672-5G
TCB4672-25GEA 360.24 CAD
TCB4672-25G TCB4672-5G
3-Bromo-2-hydroxy-5-nitropyridine ≥97.0% (by GC, titration analysis)
3-Bromo-2-hydroxy-5-nitropyridine

CAS Number: 15862-33-6
MDL Number: MFCD03840431
Molecular Formula: C5H3BrN2O3
Molecular Weight: 218.99
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 216


Other Product Names / Synonyms: 3-Bromo-5-nitro-2-pyridinol, 3-Bromo-5-nitro-2-pyridone

Formula: C5H3BrN2O3
MW: 218.99 g/mol
Melting Pt: 216 °C
MDL Number: MFCD03840431
CAS Number: 15862-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=485d498f472e1976d49b67b90ce0579a3da02e94&vid=485d498f472e1976d49b67b90ce0579a3da02e94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR