3-Bromo-4-fluorobenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (3-Bromo-4-fluorophenyl)methanol

B4656-25G B4656-5G
TCB4656-25GEA 352.34 CAD
TCB4656-25G TCB4656-5G
3-Bromo-4-fluorobenzyl alcohol ≥98.0%
3-Bromo-4-fluorobenzyl alcohol

CAS Number: 77771-03-0
MDL Number: MFCD00143093
Molecular Formula: C7H6BrFO
Molecular Weight: 205.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Specific Gravity (20/20): 1.67

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 90 °C
Density: 1.67 (20 °C)
MDL Number: MFCD00143093
CAS Number: 77771-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=399109c5fa50114730d7cb858330b520a976b52a&vid=399109c5fa50114730d7cb858330b520a976b52a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR