4-Benzyloxy-3-fluorophenylboronic acid 97.0-108.0% (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
4-Benzyloxy-3-fluorobenzeneboronic acid
B4639-1G
B4639-5G
TCB4639-1GEA
47.4
CAD
TCB4639-1G
TCB4639-5G
4-Benzyloxy-3-fluorophenylboronic acid 97.0-108.0% (contains varying amounts of Anhydride)
4-Benzyloxy-3-fluorophenylboronic acid
CAS Number: 133057-83-7
MDL Number: MFCD00994627
Molecular Formula: C13H12BFO3
Molecular Weight: 246.04
Form: Crystal
Color: White
Melting point (°C): 131
Other Product Names / Synonyms: 4-Benzyloxy-3-fluorobenzeneboronic Acid
Formula:
C₁₃H₁₂BFO₃ MW: 246.04 g/mol Melting Pt: 131 °C |
MDL Number:
MFCD00994627 CAS Number: 133057-83-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...