Benzyl acrylate ≥97.0% (by GC) stabilized

Supplier: TCI America

B4627-100G B4627-25G
TCB4627-100GEA 218.2 CAD
TCB4627-100G TCB4627-25G
Benzyl acrylate ≥97.0% (by GC) stabilized
Benzyl acrylate

(stabilized with MEHQ)
CAS Number: 2495-35-4
MDL Number: MFCD00048147
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 108
Specific Gravity (20/20): 1.06


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Benzyl Ester

Formula: C₁₀H₁₀O₂
Boiling Pt: 60 °C
Density: 1.06 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00048147
CAS Number: 2495-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c81813b15fb608155df0187d16353a7b0fbb9144&vid=c81813b15fb608155df0187d16353a7b0fbb9144&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR