(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98,0

Supplier: TCI America

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic Acid 4-tert-Butyl Ester, N-Fmoc-D-aspartic Acid 4-tert-Butyl Ester, 4-tert-Butyl N-Fmoc-D-aspartate, Fmoc-D-Asp(OtBu)-OH

B4619-1G B4619-5G
TCB4619-1GEA 34.76 CAD
TCB4619-1G TCB4619-5G
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98,0
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

CAS Number: 112883-39-3
MDL Number: MFCD00077050
Molecular Formula: C23H25NO6
Molecular Weight: 411.45
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 149
Specific rotation [a]20/D: 25 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-tert-Butyl N-Fmoc-D-aspartate, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic Acid 4-tert-Butyl Ester, N-Fmoc-D-aspartic Acid 4-tert-Butyl Ester, Fmoc-D-Asp(OtBu)-OH

Formula: C₂₃H₂₅NO₆
MW: 411.5 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00077050
CAS Number: 112883-39-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be95adba19dbd25d3ba9b3639e339b5624880575&vid=be95adba19dbd25d3ba9b3639e339b5624880575&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR