O-tert-Butylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

B4589-1G B4589-5G
TCB4589-1GEA 104.28 CAD
TCB4589-1G TCB4589-5G
O-tert-Butylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
O-tert-Butylhydroxylamine hydrochloride

CAS Number: 39684-28-1
MDL Number: MFCD00043272
Molecular Formula: C4H11NO
Molecular Weight: 125.60
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 156


Product Categories: Synthetic Organic Chemistry, Nucleophiles (Aromatic VNS Reaction)
Other Product Names / Synonyms: tert-Butoxyamine Hydrochloride

Formula: C₄H₁₂ClNO
Melting Pt: 156 °C
MDL Number: MFCD00043272
CAS Number: 39684-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afd3056173ba617381d185529ca8f2878f39f178&vid=afd3056173ba617381d185529ca8f2878f39f178&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR