7-Bromo-2-naphthol ≥97.0%
Supplier: TCI America
Synonyms:
7-Bromonaphthalen-2-ol, 7-Bromo-2-hydroxynaphthalene, 2-Bromo-7-hydroxynaphthalene
B4580-1G
B4580-5G
TCB4580-1GEA
63.2
CAD
TCB4580-1G
TCB4580-5G
7-Bromo-2-naphthol ≥97.0%
7-Bromo-2-naphthol
CAS Number: 116230-30-9
MDL Number: MFCD00277644
Molecular Formula: C10H7BrO
Molecular Weight: 223.07
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 134
Formula:
C₁₀H₇BrO MW: 223.07 Melting Pt: 134 °C |
MDL Number:
MFCD00277644 CAS Number: 116230-30-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...