2-Bromo-4-thiazolecarboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
2-Bromo-1,3-thiazole-4-carboxylic acid, 2-Bromothiazole-4-carboxylic acid
B4578-1G
B4578-5G
TCB4578-1GEA
120.08
CAD
TCB4578-1G
TCB4578-5G
2-Bromo-4-thiazolecarboxylic acid ≥98.0%
2-Bromo-4-thiazolecarboxylic acid
CAS Number: 5198-88-9
MDL Number: MFCD04115729
Molecular Formula: C4H2BrNO2S
Molecular Weight: 208.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 217
Lambda max.: 243 nm (MeOH)
Formula:
C₄H₂BrNO₂S MW: 208.04 g/mol Melting Pt: 217 °C |
MDL Number:
MFCD04115729 CAS Number: 5198-88-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...