9-(4-Biphenylyl)-10-bromoanthracene ≥98.0%

Supplier: TCI America

B4569-1G B4569-5G
TCB4569-1GEA 91.64 CAD
TCB4569-1G TCB4569-5G
9-(4-Biphenylyl)-10-bromoanthracene ≥98.0%
9-(4-Biphenylyl)-10-bromoanthracene

CAS Number: 400607-05-8
MDL Number: MFCD19440858
Molecular Formula: C26H17Br
Molecular Weight: 409.33
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 253

Formula: C₂₆H₁₇Br
Melting Pt: 253 °C
MDL Number: MFCD19440858
CAS Number: 400607-05-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b01e5c174e1dcdd7c48eb62d8d458963def2c3a&vid=8b01e5c174e1dcdd7c48eb62d8d458963def2c3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR