Benzylamine hydroiodide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B4566-1G
B4566-5G
TCB4566-1GEA
133.83
CAD
TCB4566-1G
TCB4566-5G
Benzylamine hydroiodide ≥98.0% (by HPLC, total nitrogen)
Benzylamine hydroiodide
CAS Number: 45579-91-7
Molecular Formula: C7H9N
Molecular Weight: 235.07
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 178
Formula:
C₇H₉N Melting Pt: 178 °C |
CAS Number:
45579-91-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...