Bis(pentafluorophenyl)phenylphosphine ≥98.0%

Supplier: TCI America

Synonyms: Bis(perfluorophenyl)(phenyl)phosphine, SpectraSynQ 443

B4560-1G B4560-200MG
TCB4560-1GEA 294.2 CAD
TCB4560-1G TCB4560-200MG
Bis(pentafluorophenyl)phenylphosphine ≥98.0%
Bis(pentafluorophenyl)phenylphosphine

CAS Number: 5074-71-5
MDL Number: MFCD00000291
Molecular Formula: C18H5F10P
Molecular Weight: 442.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 105
Melting point (°C): 60
Flash Point (°C): 113

Formula: C₁₈H₅F₁₀P
MW: 442.2 g/mol
Boiling Pt: 105 °C
Melting Pt: 60 °C
Flash Pt: 113 °C
MDL Number: MFCD00000291
CAS Number: 5074-71-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4574aabf005f03e8bc860f2600ab1a4064db7cf&vid=d4574aabf005f03e8bc860f2600ab1a4064db7cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR