2H-1,4-Benzothiazin-3(4H)-one ≥98.0%
Supplier: TCI America
B4554-1G
B4554-5G
B4554-25G
TCB4554-1GEA
45.82
CAD
TCB4554-1G
TCB4554-5G
TCB4554-25G
2H-1,4-Benzothiazin-3(4H)-one ≥98.0%
2H-1,4-Benzothiazin-3(4H)-one
CAS Number: 5325-20-2
MDL Number: MFCD00006698
Molecular Formula: C8H7NOS
Molecular Weight: 165.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 180
Formula:
C₈H₇NOS Melting Pt: 180 °C |
MDL Number:
MFCD00006698 CAS Number: 5325-20-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...