2H-1,4-Benzoxazin-3(4H)-one ≥98.0% (by GC)

Supplier: TCI America

B4553-25G B4553-5G
TCB4553-25GEA 110.6 CAD
TCB4553-25G TCB4553-5G
2H-1,4-Benzoxazin-3(4H)-one ≥98.0% (by GC)
2H-1,4-Benzoxazin-3(4H)-one

CAS Number: 5466-88-6
MDL Number: MFCD00158536
Molecular Formula: C8H7NO2
Molecular Weight: 149.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 174
Lambda max.: 286 nm (EtOH)

Formula: C₈H₇NO₂
Melting Pt: 174 °C
MDL Number: MFCD00158536
CAS Number: 5466-88-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=728c5e800c41c7e82f4c17d10c6004c70121998c&vid=728c5e800c41c7e82f4c17d10c6004c70121998c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR