N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4550-25G B4550-5G
TCB4550-25GEA 244.58 CAD
TCB4550-25G TCB4550-5G
N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine ≥98.0% (by HPLC, total nitrogen)
N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine

CAS Number: 499128-71-1
MDL Number: MFCD20261458
Molecular Formula: C30H22BrN
Molecular Weight: 476.42
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 171


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4-Bromo-4',4''-diphenyltriphenylamine

Formula: C₃₀H₂₂BrN
Melting Pt: 171 °C
MDL Number: MFCD20261458
CAS Number: 499128-71-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b69351e2c81b1d16f558859f15f6e6b765b9e03b&vid=b69351e2c81b1d16f558859f15f6e6b765b9e03b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR