4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene ≥98.0%

Supplier: TCI America

B4539-1G B4539-5G
TCB4539-1GEA 64.46 CAD
TCB4539-1G TCB4539-5G
4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene ≥98.0%
4-Bromo-1-methoxy-2-(3-methoxypropoxy)benzene

CAS Number: 173336-76-0
MDL Number: MFCD14582816
Molecular Formula: C11H15BrO3
Molecular Weight: 275.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 129
Melting point (°C): 36
Storage Temperature: 0-10°C

Formula: C₁₁H₁₅BrO₃
Boiling Pt: 129 °C
Melting Pt: 36 °C
MDL Number: MFCD14582816
CAS Number: 173336-76-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d310dc0fe76af9b8e9d146d3091a003bd15fb26&vid=0d310dc0fe76af9b8e9d146d3091a003bd15fb26&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR