9-(4-Biphenylyl)-3-bromocarbazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 9-([1,1'-Biphenyl]-4-yl)-3-bromo-9H-carbazole

B4538-25G B4538-5G
TCB4538-25GEA 557.11 CAD
TCB4538-25G TCB4538-5G
9-(4-Biphenylyl)-3-bromocarbazole ≥98.0% (by GC)
9-(4-Biphenylyl)-3-bromocarbazole

CAS Number: 894791-46-9
MDL Number: MFCD16038226
Molecular Formula: C24H16BrN
Molecular Weight: 398.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 155

Formula: C₂₄H₁₆BrN
MW: 398.30 g/mol
Melting Pt: 155 °C
MDL Number: MFCD16038226
CAS Number: 894791-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95921715d92c6a0e7ab229d9010cf72c137234e5&vid=95921715d92c6a0e7ab229d9010cf72c137234e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR