N,N'-Bis(tert-butoxycarbonyl)-S-methylisothiourea ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
Methyl N,N'-Bis(tert-butoxycarbonyl)carbamimidothioate, N,N'-Di-Boc-carbamimidothioic Acid Methyl Ester, 1,3-Di-Boc-2-methylisothiourea, N,N'-Bis(tert-butoxycarbonyl)carbamimidothioic Acid Methyl Ester, N,N'-Di-Boc-S-methylisothiourea, Methyl N,N'-Di-Boc-carbamimidothioate, 1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea
CAS Number: 107819-90-9
MDL Number: MFCD00239356
Molecular Formula: C12H22N2O4S
Molecular Weight: 290.38
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 123
Product Categories: Synthetic Organic Chemistry, Guanidinylation
Formula:
C₁₂H₂₂N₂O₄S MW: 290.38 g/mol Melting Pt: 123 °C |
MDL Number:
MFCD00239356 CAS Number: 107819-90-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...