3-Bromo-5-(trifluoromethyl)benzoic acid ≥98.0%
Supplier: TCI America
B4533-25G
B4533-5G
TCB4533-25GEA
535.62
CAD
TCB4533-25G
TCB4533-5G
3-Bromo-5-(trifluoromethyl)benzoic acid ≥98.0%
3-Bromo-5-(trifluoromethyl)benzoic acid
CAS Number: 328-67-6
MDL Number: MFCD03412186
Molecular Formula: C8H4BrF3O2
Molecular Weight: 269.02
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 135
Formula:
C₈H₄BrF₃O₂ MW: 269.02 g/mol Melting Pt: 135 °C |
MDL Number:
MFCD03412186 CAS Number: 328-67-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...